Skip to Content
Merck
All Photos(1)

Key Documents

31707

Supelco

Dimetridazole

VETRANAL®, analytical standard

Synonym(s):

1,2-Dimethyl-5-nitroimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O2
CAS Number:
Molecular Weight:
141.13
Beilstein:
130665
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ncc(n1C)[N+]([O-])=O

InChI

1S/C5H7N3O2/c1-4-6-3-5(7(4)2)8(9)10/h3H,1-2H3

InChI key

IBXPYPUJPLLOIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimetridazole (DMZ) is a 5-nitroimidazole type veterinary drug mostly used for treating infections caused by bacteria and protozoa. These infections can be frequently seen in the poultry industry.

Application

Dimetridazole has been used as reference standard for determination of DMZ concentration from poultry muscles, porcine kidney and liver, and chicken liver tissues using gas chromatography–electron capture negative ionization mass spectrometry (GC–ECNI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of dimetridazole and metronidazole in poultry and porcine tissues by gas chromatography?electron capture negative ionization mass spectrometry.
Ho, Clare, et al.
Analytica Chimica Acta, 530.1, 23-31 (2005)
J H Wang
Journal of chromatography. A, 918(2), 435-438 (2001-06-16)
A method was developed for the determination of the nitroimidazole compounds dimetridazole (DMZ), ronidazole (RNZ) and metronidazole (MNZ) by gas chromatography with nitrogen phosphorus detection. Nitroimidazole compounds were extracted with acetonitrile, followed by acidification using acetic acid and cleanup using
M G Müller et al.
Veterinary parasitology, 152(1-2), 67-78 (2008-01-02)
Four falcons from a private collection of 137 falcons in Abu Dhabi (UAE) died suddenly in summer 2005. In order to screen for a possible disease among the remaining falcons in the aviary, all other birds were caught, examined and
M-P Callait-Cardinal et al.
The Veterinary record, 161(17), 581-585 (2007-10-30)
Between April 2003 and March 2005, 113 outbreaks of histomonosis were recorded in standard turkey farms in France, and 15 cases were recorded in turkey breeding centres. Most of the cases were in north-west France, the principal farming area for
Jianzhong Shen et al.
Journal of AOAC International, 86(3), 505-509 (2003-07-11)
A liquid chromatographic method was developed for determination of metronidazole (MNZ), ronidazole (RNZ), dimetridazole (DMZ), and 2-hydroxymethyl-1-methyl-5-nitroimidazole (DMZOH) in swine tissue. After extraction with ethyl acetate and evaporation, the nitroimidazoles were redissolved in hydrochloric acid. Hexane was used in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service