Skip to Content
Merck
All Photos(2)

Key Documents

211982

Sigma-Aldrich

Solvent Green 3

Dye content 95 %

Synonym(s):

D & C Green No. 6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22N2O2
CAS Number:
Molecular Weight:
418.49
Colour Index Number:
61565
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 95%

color

violet

mp

220-221 °C (lit.)

solubility

pyridine: 1%

λmax

644-607 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(Nc2ccc(Nc3ccc(C)cc3)c4C(=O)c5ccccc5C(=O)c24)cc1

InChI

1S/C28H22N2O2/c1-17-7-11-19(12-8-17)29-23-15-16-24(30-20-13-9-18(2)10-14-20)26-25(23)27(31)21-5-3-4-6-22(21)28(26)32/h3-16,29-30H,1-2H3

InChI key

TVRGPOFMYCMNRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Solvent Green 3, also known as Quinizarine Green SS (CI 61565) is an anthraquinone derivative. It is a green dye that imparts its color to materials. Solvent Green 3 is made by condensing either leucoquinizarin or a mixture of leucoquinizarin and quinizarin with two moles of p-toluidine in the presence of boric acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryam Mohamadi et al.
Journal of AOAC International, 94(3), 959-967 (2011-07-30)
Dispersive liquid-liquid microextraction (DLLME) based on a task-specific ionic liquid (TSIL) was developed for the extraction and preconcentration of trace amounts of cadmium from aqueous samples, followed by flame atomic absorption spectrometry (FAAS) determination. In the proposed approach, cadmium ions
Kazunori Iwanaga et al.
Biological & pharmaceutical bulletin, 29(3), 508-512 (2006-03-02)
Solvent Green 3 (SG), a model poorly water-soluble compound, was orally administered to rats with soybean oil emulsion or the Self-microemulsifying drug delivery system (SMEDDS) composed of Gelucire44/14. The bioavailability of SG after oral administration with SMEDDS was 1.7-fold higher
Yoshiaki Ikarashi et al.
Kokuritsu Iyakuhin Shokuhin Eisei Kenkyujo hokoku = Bulletin of National Institute of Health Sciences, (123)(123), 53-56 (2006-03-18)
The cytotoxicities of chemicals used in household products were evaluated using a neutral red (NR) uptake assay. The chemicals tested during 1997-2004 were rubber additives (accelerators, antioxidants and retarders), solvents, plasticizers and biocides, such as antimicrobials, fungicides, preservatives used in
Does subcuticular green polyglycolic acid suture tattoo?
L L Reti et al.
The Australian & New Zealand journal of obstetrics & gynaecology, 22(3), 153-154 (1982-08-01)
Liquid chromatographic separations of basic compounds.
David V McCalley
Advances in chromatography, 46, 305-350 (2008-11-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service