Skip to Content
Merck
All Photos(1)

Key Documents

09772

Sigma-Aldrich

Aluminum silicate

synthetic, 82% SiO2 basis (based on calcined substance), 9.5% Al2O3 basis (based on calcined substance), 8% Na2O basis (based on calcined substance)

Synonym(s):

Aluminum sodium silicate, P 820 A

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

composition

Al2O3, 9.5% (based on calcined substance)
Na2O, 8% (based on calcined substance)
SiO2, 82% (based on calcined substance)

loss

≤18% loss on ignition

InChI

1S/Al.Na.2O3Si/c;;2*1-4(2)3/q+3;+1;2*-2

InChI key

URGAHOPLAPQHLN-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Roland et al.
Poultry science, 70(4), 805-811 (1991-04-11)
Experiments were conducted to determine if the high ion-exchange capacity of sodium aluminosilicate (ZA) marketed as ETHACAL Feed Component or its aluminum content or both are related to the beneficial effect of ZA on egg specific gravity (ESG). In Experiments

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service