Skip to Content
Merck
All Photos(1)

Key Documents

11965085001

Roche

Anti-His6-Peroxidase

from mouse IgG1

Synonym(s):

antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BMG-his-1, monoclonal

form

lyophilized

packaging

pkg of 50 U

manufacturer/tradename

Roche

isotype

IgG1

storage temp.

2-8°C

General description

Anti-His6-Peroxidase is a monoclonal antibody to His6-tagged proteins, conjugated to horseradish peroxidase. Anti-His6-Peroxidase specifically recognizes an epitope of six consecutive histidine residues of both natural and recombinant proteins. The antibody reacts with native and denatured histidine-tagged fusion proteins independent of the epitope-sequence location; however, it preferentially recognizes the C-terminal His6 epitope with high sensitivity.

Specificity

Anti-His6-Peroxidase specifically recognizes an epitope of six consecutive histidine residues (His6) in natural and recombinant proteins. It reacts with both native and denatured histidine-tagged fusion proteins, independent of location of the epitopesequence. However, Anti-His6-Peroxidase preferentially recognizes the C-terminal His6 epitope with high sensitivity. Anti-His6 allows specific and sensitive detection of histidine-tagged proteins irrespective of the expression system used.
The antibody recognizes an epitope of six consecutive histidine residues (His6) in natural and recombinant proteins. It reacts with both native and denatured histidine-tagged fusion proteins, independent of the epitope-sequence location.

Application

Anti-His6-Peroxidase has been used in in vitro serum stability of fusion toxins, pull-down assay and western blotting.
Anti-His6-Peroxidase is used for the detection of His6-tagged proteins in:
  • ELISA (enzyme-linked immunosorbent assay)
  • Western blot

Quality

Function test: Western blot using extracts from cell line expressing a recombinant His6-tagged protein.

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate.


The following concentrations should be taken as a guideline:
  • ELISA: 100 mU/ml
  • Western blot: 100mU/ml

Reconstitution

Add 1 ml double-distilled water to a final concentration of 50 U/ml.
Reconstitution should be performed for at least 10 minutes.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaheh Gheybi et al.
Cancer reports (Hoboken, N.J.), 6(3), e1745-e1745 (2022-10-28)
CD44, as a tumor-associated marker, can be used to detect stem cells in breast cancer. While CD44 is expressed in normal epithelial cells, carcinoma cells overexpress CD44. In the current study, we designed a recombinant protein that included the variable
KaiC intersubunit communication facilitates robustness of circadian rhythms in cyanobacteria
Kitayama Y, et al.
Nature Communications, 4 (2013)
Andra-Octavia Roman et al.
Nature communications, 13(1), 876-876 (2022-02-17)
The membrane receptor kinases HAESA and HSL2 recognize a family of IDA/IDL signaling peptides to control cell separation processes in different plant organs. The homologous HSL1 has been reported to regulate epidermal cell patterning by interacting with a different class
Identification and Characterization of Heptaprenylglyceryl Phosphate Processing Enzymes in Bacillus subtilis
Linde M, et al.
The Journal of Biological Chemistry, 291(28), 14861-14870 (2016)
Clare Sheridan et al.
Molecular cell, 31(4), 570-585 (2008-08-30)
Bax and Bak promote apoptosis by perturbing the permeability of the mitochondrial outer membrane and facilitating the release of cytochrome c by a mechanism that is still poorly defined. During apoptosis, Bax and Bak also promote fragmentation of the mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service