SHF1350425
Hi-Flow™ Plus Membrane 135
backing thickness 4 mil, width 25 mm, pack of 1 Slit roll
Synonym(s):
HF, Hi-Flow, Hi-Flow™ Plus HF13504, immunoassay membrane, lateral flow membrane, nitrocellulose membrane
About This Item
Recommended Products
packaging
pack of 1 Slit roll
Quality Level
parameter
135 sec/4 cm capillary flow rate
backing thickness
4 mil
roll L
100 m (328 ft)
width
25 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Hi-Flow™ Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing and animal health.
- Hi-Flow™ Plus membranes are suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
- They are optimised for use in both qualitative and quantitative test types and pair well with lateral flow detector particles including colloidal gold nanoparticles, dyed/ fluorescent microspheres etc.
- Hi-Flow™ Plus membranes are suitable for use in lateral flow tests designed to detect analytes present in a broad range of samples matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat and others.
Features and Benefits
- Consistent Flow & Surface Chemistry, Robust manufacturing process
- Proprietary Surface Chemistry Optimised for Lateral Flow Applications
- High Quality Level: MQ400
- Trusted Partner and Brand with Decades of membrane manufacturing experience
- High production capacity, high stock levels, multiple manufacturing lines
- Convenient samples packs available in several formats
- Manufacturing scale quantities available to purchase online with immediate shipping.
- Consistent capillary flow time and membrane thickness
- Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface
- Sharp, intense, and uniform test and control line formation
- Excellent intra-lot and inter-lot consistency.
- Better Assay sensitivity
- Excellent reproducibility
- Improved lateral flow test manufacturing yields.
- Quick Delivery and Lead time.
- No supply disruption
- Excellent Technical support
Other Notes
Individually packed single slit roll for sampling / evaluation
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XIV (Authorisation List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service