Skip to Content
Merck
All Photos(1)

Key Documents

RECYMAG65PMX27BK

Millipore

MILLIPLEX® Rat Cytokine/Chemokine Magnetic Bead Panel - Premixed 27 Plex Space Saver (Bulk) Packaging

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in rat serum, plasma and cell culture samples.

Synonym(s):

luminex rat cytokine chemokine growth factor multiplex assay, luminex rat pro inflammatory anti inflammatory cytokine panel, millipore rat inflammation multiplex kit, rat interleukin immunoassay panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

rat

manufacturer/tradename

Milliplex®

assay range

standard curve range: 0.2-1,000 pg/mL
(EGF)

standard curve range: 1.2-5,000 pg/mL
(GRO/KC)

standard curve range: 12.2-50,000 pg/mL
(GM-CSF, IL-1α, IL-2, IL-12 (p70), IL-18)

standard curve range: 14.6-60,000 pg/mL
(IFNγ, MIP-1α)

standard curve range: 2.4-10,000 pg/mL
(Fractalkine, IP-10, Leptin, TNFα)

standard curve range: 24.4-100,000 pg/mL
(LIX, MIP-2)

standard curve range: 4.9-20,000 pg/mL
(Eotaxin, C-CSF, IL-4, IL-5, IL-13, RANTES, VEGF)

standard curve range: 7.3-30,000 pg/mL
(IL-10, IL-17A, MCP-1)

standard curve range: 73.2-300,000 pg/mL
(IL-6)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MILLIPLEX® Rat Cytokine/Chemokine Bead Panel (RECYTMAG-65K & RECYMAG65K27PMX) enables you to focus on the therapeutic potential of cytokines, as well as the modulation of cytokine expression.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There is no or minimum cross-reactivity between the antibodies and any other analytes in the panel.

Application

  • Analytes: EGF, Eotaxin/CCL11, Fractalkine, G-CSF, GM-CSF, GRO/KC, IFN-γ, IL-1α, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12 (p70), IL-13, IL-17A, IL-18, IP-10, Leptin, LIX, MCP-1, MIP-1α, MIP-2, RANTES, TNF-α, VEGF
  • Recommended Sample type: rat serum or plasma
  • Recommended Sample dilution: 1:2
  • Assay Run Time: One day

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoxuan Chen et al.
Environmental science & technology, 54(16), 10227-10236 (2020-07-15)
Uncertainties regarding optimized air pollution control remain as the underlying mechanisms of city-specific ambient particulate matter (PM)-induced health effects are unknown. Here, water-soluble extracts of PMs collected from four global cities via automobile air-conditioning filters were consecutively injected three times
Natàlia de la Oliva et al.
Scientific reports, 8(1), 5965-5965 (2018-04-15)
Parylene C is a highly flexible polymer used in several biomedical implants. Since previous studies have reported valuable biocompatible and manufacturing characteristics for brain and intraneural implants, we tested its suitability as a substrate for peripheral nerve electrodes. We evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service