Skip to Content
Merck
All Photos(1)

Key Documents

PX1815

Supelco

n-Propyl Alcohol

GR

Synonym(s):

n-Propyl Alcohol, 1-Propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-746-9
bp:
96.5-98 °C/1013 hPa
vapor pressure:
19 hPa ( 20 °C)

vapor pressure

19 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

evapn. residue

≤0.01%

color

APHA: ≤10

pH

7 (20 °C, 200 g/L in H2O)

bp

96.5-98 °C/1013 hPa

mp

-127 °C

transition temp

flash point 15 °C

density

0.8 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCCC

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Application

GR

Analysis Note

Assay (GC): 98.0% min
Color (APHA): 10 max
Evaporation residue: 0.01% max
Form: Clear liquid

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service