Skip to Content
Merck
All Photos(2)

Documents

MABN774

Sigma-Aldrich

Anti-TDP-43 Antibody, clone DB9

clone DB9, from mouse

Synonym(s):

TAR DNA-binding protein 43, TDP-43, TDP43, TAR DNA binding protein-43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DB9, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TARDBP(23435)

General description

TAR DNA-binding protein 43 (TDP-43) belongs to the hnRNP protein family and plays an important role in transcription, pre-mRNA splicing, mRNA stability and mRNA transport. It is involved in splicing of the apolipoprotein A-II and cystic fibrosis transmembrane gene. This protein is highly expressed in the pancreas, placenta, lung, genital tract and spleen. Mutations in TDP-43 have been associated with amyotrophic lateral sclerosis, frontotemporal dementia, Parkinson′s disease and Alzheimer′s disease.

Immunogen

The antibody was raised against an immunogen consisting of His-tagged recombinant protein corresponding to amino acid residues 208-414 of human TDP-43.

Application

Detect TDP-43 using this Anti-TDP-43 Antibody, clone DB9 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TDP-43 in human cerebellum, pancreas, and thalamus tissues.

Quality

Evaluated by Western Blotting in SH-SY-5Y cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected TDP-43 in 10 µg of SH-SY-5Y cell lysate.

Target description

~43 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Silva-Hucha et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 21(3), e00340-e00340 (2024-03-13)
Amyotrophic lateral sclerosis (ALS) is a rare neuromuscular disease characterized by severe muscle weakness mainly due to degeneration and death of motor neurons. A peculiarity of the neurodegenerative processes is the variable susceptibility among distinct neuronal populations, exemplified by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service