Skip to Content
Merck
All Photos(1)

Key Documents

MAB3507

Sigma-Aldrich

Anti-Caspase 2 Antibody, clone 11B4

clone 11B4, Chemicon®, from rat

Synonym(s):

ICH-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

11B4, monoclonal

species reactivity

mouse, monkey, human, canine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP2(835)

Related Categories

Specificity

MAB3507 antibody recognizes a reactive epitope that lies in the P19 subunit of human Caspase-2 (ICH-1, Nedd-2).

Immunogen

Raised against the P19 subunit of recombinant human Caspase-2 protein.

Application

Anti-Caspase 2 Antibody, clone 11B4 is an antibody against Caspase 2 for use in ELISA, FC, IP & WB.
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
Western blot: Recommended usage at 5 μg/ml, which has been found to detect endogenous Caspase-2 in 5 x 10E5 HeLa and MCF-7 cells.

Immunoprecipitation: Recommended concentration 1-5 μg/ml FACS: Recommended concentration 1 μg/ml. It is necessary to permeabilize the cell membrane prior to analysis.

EIA: Recommended concentration 0.5-1 μg/ml.

Immunohistochemistry: Use at 10-20 μg/ml. It is necessary to fix the cells in 4% Paraformaldehyde/0.18% Triton-X100 for 10 minutes prior to staining.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-571

Physical form

Format: Purified
Liquid with 0.05% Bronidox as a preservative.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Del Nagro et al.
Cell cycle (Georgetown, Tex.), 13(2), 303-314 (2013-11-20)
Activation of Checkpoint kinase 1 (Chk1) following DNA damage mediates cell cycle arrest to prevent cells with damaged DNA from entering mitosis. Here we provide a high-resolution analysis of cells as they undergo S- and G₂-checkpoint bypass in response to
Brittany L Carroll et al.
FEBS open bio, 8(1), 27-40 (2018-01-13)
Sphingosine kinase 1 (SK1) is a lipid kinase whose activity produces sphingosine 1-phosphate, a prosurvival lipid associated with proliferation, angiogenesis, and invasion. SK1 overexpression has been observed in numerous cancers. Recent studies have demonstrated that SK1 proteolysis occurs downstream of
Ramaswamy Sharma et al.
PloS one, 9(4), e93696-e93696 (2014-04-03)
Osteoporosis is a silent disease, characterized by a porous bone micro-structure that enhances risk for fractures and associated disabilities. Senile, or age-related osteoporosis (SO), affects both men and women, resulting in increased morbidity and mortality. However, cellular and molecular mechanisms
M J Parsons et al.
Cell death and differentiation, 20(9), 1174-1182 (2013-05-07)
Despite being the most evolutionarily conserved of the mammalian caspases, little is understood about the cellular function of caspase-2 in normal tissues or what role caspase-2 may have in the progression of human disease. It has been reported that deletion
Lauren T Evans et al.
The EMBO journal, 40(4), e105106-e105106 (2020-12-23)
Centriole copy number is tightly maintained by the once-per-cycle duplication of these organelles. Centrioles constitute the core of centrosomes, which organize the microtubule cytoskeleton and form the poles of the mitotic spindle. Centrosome amplification is frequently observed in tumors, where

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service