Skip to Content
Merck
All Photos(1)

Key Documents

MAB13458

Sigma-Aldrich

Anti-MMP-9 Antibody, clone 9D4.2

clone 9D4.2, Chemicon®, from mouse

Synonym(s):

Gelatinase B, 92 kDa Type IV Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9D4.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

Specificity

Human MMP-9 pro and active forms.

Immunogen

Purified Human MMP-9 (Gelatinase B)

Application

Detect MMP-9 using this Anti-MMP-9 Antibody, clone 9D4.2 validated for use in ELISA & WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western blot: 1:200- 1:1000

Immunocytochemistry: Not Tested

ELISA: 1:200-1:1000

Immunohistochemistry: Not Tested

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-1150

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.01% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6/12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter Maetzler et al.
PloS one, 11(3), e0149349-e0149349 (2016-03-05)
Based on animal and ex-vivo experiments, Growth/Differentiation Factor-15 (GDF15, also called Macrophage Inhibitory Cytokine-1, MIC1), a member of the transforming growth factor-beta family, and Matrix Metalloproteinase-9 (MMP9), a member of the matrix metalloprotease family may be potential markers for Lewy
Prity Sahay et al.
Investigative ophthalmology & visual science, 58(6), BIO106-BIO113 (2017-06-07)
To evaluate the differential expression of tear matrix metalloproteinases (MMP) 2 and 9 in of patients with various forms of glaucoma. Tear samples were collected with a Schirmer's strip from 148 eyes of 113 patients (medically naïve patients with primary
Prity Sahay et al.
PloS one, 16(4), e0249759-e0249759 (2021-04-30)
Pseudoexfoliation (PXF) is a unique form of glaucoma characterized by accumulation of exfoliative material in the eyes. Changes in tear profile in disease stages may give us insights into molecular mechanisms involved in causing glaucoma in the eye. All patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service