Skip to Content
Merck
All Photos(2)

Key Documents

HGAMMAG-303E

Millipore

MILLIPLEX® Human Immunoglobulin IgE Single Plex Magnetic Bead Kit - Isotyping Assay

Isotyping Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple immunoglobulins (Ig) in human serum, plasma and cell culture samples.

Synonym(s):

Human IgE Bead Kit, Human IgE Detection Kit, Immunoglobulin IgE Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 102%
(IgE)

sensitivity: 0.4 ng/mL
standard curve range: 4.1-1,000 ng/mL
(IgE)

technique(s)

multiplexing: suitable

compatibility

configured for Single Plex

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Found only in mammals, Immunoglobulin E (IgE) plays an essential role in allergic reactions and is predominantly associated with type 1 hypersensitivity. IgE is the least abundant immunoglobulin isotype in normal individuals, present at only 0.05% of IgG serum concentrations. It is capable of triggering robust immune reactions, with up to 10 times the typical level detected in the serum of atopic patients, although increased levels occur only in approximately 50% of patients with allergic disorders. IgE binds with high affinity to FcεRI, found primarily on mast cells and basophils, priming the cells to recognize specific antigens such as dust mites, pet dander or pollen. Once primed and with repeated exposure to the same antigen, these cells release molecules such as histamines, prostaglandins, and leukotrienes, resulting in an “allergic reaction”.

Although the mechanisms of this immune response are well understood, continued research is needed to understand the varying degrees of allergic responses – from mild rhinitis to anaphylaxis. Consequently, the MILLIPLEX® Human IgE Bead Single Plex Kit enables you to measure levels of IgE in only 50 μL of 1:50 diluted human serum or plasma, saving both time and precious sample.

Panel Type: Immune Response

Specificity

Cross-reactivity between these antibodies and those against other immunoglobulins is non-detectable or negligible.

Application

  • Analyte: IgE
  • Recommended Sample type: Serum, plasma, or tissue/cell lysate and culture supernatant samples
  • Recommended Sample dilution: 1:50 diluted plasma or serum, or 50 μL cell culture supernatant
  • Assay Run Time: One day
  • Research Category: Inflammation & Immunology

Packaging

96 Well Plate

Components

MILLIPLEX MAP Anti-Human k and λ Light Chain, PE

MILLIPLEX MAP Human IgE Standard

MILLIPLEX MAP Human Serum Positive Control

MILLIPLEX MAP Assay Buffer

MILLIPLEX MAP Washer Buffer, 10X

Set of one 96-Well Filter Plate with 4 Sealers

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophia Kwon et al.
Annals of the American Thoracic Society, 13(8), 1253-1261 (2016-04-21)
The World Trade Center (WTC) collapse generated caustic airborne particulates that caused chronic rhinosinusitis in exposed Fire Department of New York firefighters. Surgery was performed when symptoms remained uncontrolled despite medical management. To identify predictors of surgical intervention for chronic
José Carlos Zarzuela et al.
Frontiers in immunology, 13, 911022-911022 (2022-08-09)
Conjunctival intraepithelial lymphocytes, tear soluble molecules and commensal microbiota have important roles in the ocular mucosal immune response in healthy and diseased subjects. For the purpose of this study, the cellular and microbial populations of the conjunctiva and the lacrimal

Related Content

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service