Skip to Content
Merck
All Photos(1)

Key Documents

F3DCAL-K25

Millipore

FLEXMAP 3D Calibration Kit, 25 Uses

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
HB.01

product line

Milliplex®

technique(s)

multiplexing: suitable

General description

All xMAP Instruments requires regular calibration and performance verification testing, to ensure the system is operating correctly and to maintain data accuracy. The FLEXMAP 3D instrument is calibrated using a calibration and a verification kit, each providing 25 uses.

Application

For use with FLEXMAP 3D system®

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

FLEXMAP 3D system is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service