Skip to Content
Merck
All Photos(1)

Key Documents

ECM440

Sigma-Aldrich

β Integrin Screening Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

species reactivity

human

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

dry ice

General description

Chemicon Investigator Kits for Integrin Research are a cost-effective source of reagents for all investigators involved with this expanding field of study. The b Integrin Screening Investigator Kit is designed for the screening of beta integrin family members.

Integrins belong to a large family of cell surface proteins commonly referred to as glycoproteins. Integrins are composed of noncovalently associated alpha and beta subunits which make up heterodimers: the larger termed alpha chain and the smaller beta chain. At least 16 alpha subunits and 8 beta subunits are expressed, which can give rise to 22 different integrins. Integrins are thought to play critical roles in cell migration, differentiation, and survival. There is now considerable evidence that these transmembrane receptors play an important role in the regulation of gene expression and signal transduction. Excellent reviews of integrin structure, function, and ligand specificity may be found in: Hynes, R. 1992. Cell 69:11-25; Kramer, R. 1993. "Integrin Structure and Ligand Specificity in Cell-Matrix Interactions" in Molecular and Cellular Aspects of Basement Membranes. Academic Press.

Chemicon offers the Beta Integrin Screening Investigator Kit for studies of human beta integrin subunits. All kit reagents are also available in larger sizes. The reagents in this kit are not application specific but rather may be used in a variety of procedures, depending upon the needs of the individual investigator.

.

Application

Research Category
Cell Structure
Research Sub Category
Integrins

Components

beta1 Mouse Monoclonal Antibody (Part No. MAB1951Z-20)- 1 tube, 20 μg.

beta2 Mouse Monoclonal Antibody (Part No. MAB1962Z-20)- 1 tube, 20 μg.

beta3 Mouse Monoclonal Antibody (Part No. MAB1957Z-20)- 1 tube, 20 μg.

beta4 Mouse Monoclonal Antibody (Part No. MAB2060-20)- 1 tube, 20 μg.

beta5 Rabbit Polyclonal Antibody (Part No. AB1926-20)- 1 tube, 20 μL.

Physical form

Chemicon′s Beta Integrin Screening Kit (ECM440) contains monoclonals to beta1, beta2, beta3, beta4, and polyclonal beta5. 20 μg of each monoclonal and 20 μL of polyclonal is included in this economical kit. All components may also be purchased in larger sizes.

Storage and Stability

Store antibodies at -20°C.

Important Note: Due to the small volumes contained within the kit, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge before opening to dislodge any liquid entrapped in the container′s cap during shipment.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harry Ma et al.
Circulation research, 93(11), 1066-1073 (2003-10-25)
The migration of vascular smooth muscle cells (SMCs) from the media into the neointima and their subsequent proliferation is important in the pathogenesis of atherosclerosis. This process is regulated by multiple factors, including growth factors, and involves changes in the
Jessica Ellen Frith et al.
Stem cells and development, 21(13), 2442-2456 (2012-03-30)
Integrins provide the primary link between mesenchymal stem cells (MSCs) and their surrounding extracellular matrix (ECM), with different integrin pairs having specificity for different ECM molecules or peptide sequences contained within them. It is widely acknowledged that the type of
Identification of extracellular matrix components and their integrin receptors in the human fetal adrenal gland.
E Chamoux, L Bolduc, J G Lehoux, N Gallo-Payet
The Journal of clinical endocrinology and metabolism null
Minhong Shen et al.
Cancer cell, 35(1), 64-80 (2019-01-08)
Triple-negative breast cancer (TNBC) patients have the worst prognosis and distant metastasis-free survival among all major subtypes of breast cancer. The poor clinical outlook is further exacerbated by a lack of effective targeted therapies for TNBC. Here we show that
Jessica E Frith et al.
Journal of cell science, 125(Pt 2), 317-327 (2012-01-18)
Mesenchymal stem cells (MSCs) have attracted great interest in recent years for tissue engineering and regenerative medicine applications due to their ease of isolation and multipotent differentiation capacity. In the past, MSC research has focussed on the effects of soluble

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service