Skip to Content
Merck
All Photos(2)

Key Documents

ABN1023

Sigma-Aldrich

Anti-LILRB2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leukocyte immunoglobulin-like receptor subfamily B member 2, LIR-2, Leukocyte immunoglobulin-like receptor 2, CD85 antigen-like family member D, Immunoglobulin-like transcript 4, ILT-4, Monocyte/macrophage immunoglobulin-like receptor 10, MIR-10, CD85d,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:
1
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LILRB2(10288)

General description

Leukocyte immunoglobulin-like receptor subfamily B member 2 (UniProt Q8N423; also known as CD85 antigen-like family member D, CD85d, Ig-like transcript 4, ILT-4, Leukocyte immunoglobulin-like receptor 2, LIR-2, MIR-10, Monocyte/macrophage immunoglobulin-like receptor 10) is encoded by the LILRB2 (also known as CD85D, ILT4, LIR2, MIR10) gene (Entrez Gene ID 10288) in human. CD85d belongs to the B class of leukocyte immunoglobulin-like (LIR) receptors family, members of which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). Cd85d is expressed on immune cells and negatively regulates immune response by engaging class I MHC molecules (HLA-A, HLA-B, HLA-C) on antigen-presenting cells.

Specificity

Reacts with all 4 known human CD85d isoforms.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human LILRB2.

Application

Anti-LILRB2 Antibody is an antibody against LILRB2 for use in Western Blotting, Immunohistochemistry .
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected LILRB2 in human ovarian cancer tumor cells, mouse cerebral cortex, human brain cancer tumor cells, mouse hippocampus, and Non-Hodgkin lymphoma cells.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected LILRB2 in 10 µg of Jurkat cell lysate.

Target description

~64/120 kDa observed. The calculated molecular weight of the four isoforms range from ~52 to ~65 kDa, a band with apparent molecular weight above ~110 kDa is often seen on blots as a result of glycosylation.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacqueline G O'Leary et al.
Transplantation direct, 9(8), e1500-e1500 (2023-07-17)
Liver allografts protect renal allografts from the same donor from some, but not all, preformed donor specific alloantibodies (DSA). However, the precise mechanisms of protection and the potential for more subtle alterations/injuries within the grafts resulting from DSA interactions require

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service