Skip to Content
Merck
All Photos(3)

Key Documents

AB5682

Sigma-Aldrich

Anti-Neurogenin-2 Antibody

Chemicon®, from rabbit

Synonym(s):

MATH4a (ms), ATH 4a, Atoh4, Ngn2, Atonal Protein Homolog 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NEUROG2(63973)

General description

Neurogenin 2 (NGN2) is a helix-loop-helix class of transcription factor. Transcription factors with bHLH motifs modulate critical events in the development of the mammalian neocortex. The transition from proliferation to neurogenesis involves a coordinate increase in the activity of proneural bHLH factors, including Neurogenin 2 and a decrease in the activity of Hes and Id factors. bHLH factors have key roles in corticogenesis, affecting the timing of differentiation and the specification of cell fate. Neurog2 controls neurogenesis in the embryonic cerebral cortex directly induces the expression of the small GTP-binding protein Rnd2 in newly generated mouse cortical neurons before they initiate migration.

Specificity

Recognizes Neurogenin 2 (MATH4a), a Helix-loop-helix class of transcription factor.

Immunogen

Synthetic peptide, amino acids 85-97 of mouse Neurogenin 2.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neuronal & Glial Markers
This Anti-Neurogenin-2 Antibody is validated for use in ELISA, IC, IH, IH(P), WB for the detection of Neurogenin-2.
Western blot: 1-10 μg/mL using ECL.
Immunocytochemistry: Cat. # AB5682 detected Neurogenin-2 in PC12 cells at 1:500 dilution. It was also detected in paraformaldehyde-fixed differentiated human neural stem cells.
ELISA: 0.5-1 μg/mL using 1 mg/mL control peptide (Catalog Number AG352) per well.
Immunohistochemistry on paraffin sections has been reported by Lamolet, B. et al. (2004)
Optimal working dilutions must be determined by the end user.

Target description

~28 kDa

Linkage

Replaces: AB9620

Physical form

Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Stable for 1 year at -70ºC from date of receipt.

Analysis Note

Control
Human brain Lysate.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan-Ci Chen et al.
Journal of neuroinflammation, 16(1), 187-187 (2019-10-14)
The aryl hydrocarbon receptor (AHR) is a ligand-dependent transcription factor activated by environmental agonists and dietary tryptophan metabolites for the immune response and cell cycle regulation. Emerging evidence suggests that AHR activation after acute stroke may play a role in
Hongwon Kim et al.
Stem cell reports, 12(3), 518-531 (2019-02-26)
Recent advances in generating three-dimensional (3D) organoid systems from stem cells offer new possibilities for disease modeling and drug screening because organoids can recapitulate aspects of in vivo architecture and physiology. In this study, we generate isogenic 3D midbrain organoids with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service