8.43945
Ammonium tetrafluoroborate
for synthesis
Synonym(s):
Ammonium tetrafluoroborate
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (acidimetric)
form
powder
pH
3.5 (20 °C, 5 g/L in H2O)
density
1.85 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
[B+3]([F-])([F-])([F-])[F-].[N+H4]
InChI
1S/BF4.H3N/c2-1(3,4)5;/h;1H3/q-1;/p+1
InChI key
PDTKOBRZPAIMRD-UHFFFAOYSA-O
Analysis Note
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service