Skip to Content
Merck
All Photos(1)

Key Documents

8.43871

Sigma-Aldrich

Polyvinyl alcohol, partially hydrolyzed

(Mw approx.200000) for synthesis

Synonym(s):

Polyvinyl alcohol, partially hydrolyzed, PVA, PVOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H6O2 · C2H4O)n
CAS Number:
MDL number:
UNSPSC Code:
13111042
NACRES:
NA.22

Quality Level

form

solid

autoignition temp.

>230 °C

mp

>200 °C

density

1.3 g/cm3 at 20 °C

bulk density

400‑600 kg/m3

storage temp.

2-30°C

Analysis Note

Degree of hydrolysis (calculated on dried substance): ≥ 85.0 %
dyn.Viscosity, 20°C (4 %; water): 34.0 - 46.0 mPa∙s
Loss on drying (110 °C 2h): ≤ 5.0 %
Ash: ≤ 1.0 %
Ester value: 130 - 150

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service