8.42649
Gallic acid
anhydrous, for synthesis
Synonym(s):
Gallic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6O5
CAS Number:
Molecular Weight:
170.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-749-9
NACRES:
NA.22
grade
anhydrous
Quality Level
Assay
≥98.0% (acidimetric)
storage temp.
2-30°C
SMILES string
Oc1c(cc(cc1O)C(=O)O)O
InChI
1S/C7H6O5/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,8-10H,(H,11,12)
InChI key
LNTHITQWFMADLM-UHFFFAOYSA-N
General description
(anhydrous) for synthesis
Application
- Gallic acid in tumor-targeted therapy: A study described the use of gallic acid-loaded HFZIF-8 nanoparticles for enhanced tumor-targeted delivery and thermal-catalytic therapy, showcasing potential applications in cancer treatment strategies (Yang et al., 2024).
- Gallic acid in industrial processes: Research highlighted the integration of gallic acid within porous sponges for efficient dye adsorption and emulsion separation, indicating its utility in environmental and industrial applications (Yan et al., 2024).
- Gallic acid in extraction processes: The use of gallic acid was explored in the ultrasound-assisted extraction of bioactive pigments from Spirulina platensis, employing natural deep eutectic solvents to enhance extraction efficiency and safety (Martins et al., 2023).
- Gallic acid in self-healing materials: A novel study developed a self-healing hydrogel using gallic acid and carboxymethyl cellulose, triggered by Fe(3+) coordination cross-linking, demonstrating innovative applications in materials science (Yang et al., 2024).
Analysis Note
Assay (acidimetric): ≥ 98.0 %
Identity (IR): passes test
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service