Skip to Content
Merck
All Photos(1)

Key Documents

8.20876

Sigma-Aldrich

Nickel catalyst

(Produced according to a process of the type developed by Murray Raney, activated for hydrogenation) for synthesis

Synonym(s):

Nickel catalyst acc. Raney

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NA.22

Quality Level

form

suspension

reaction suitability

reagent type: reductant

pH

9-11 (20 °C in H2O, suspension)

density

>1.2 g/cm3

storage temp.

2-30°C

Application

  • Bifunctional Electrocatalysis: Suresh et al. developed multi-active sites loaded NiCu-MOF@MWCNTs as a bifunctional electrocatalyst for water splitting, demonstrating the potential of nickel catalysts in enhancing both hydrogen and oxygen evolution reactions, thereby offering a robust solution for electrochemical energy conversion (Suresh et al., 2024).

Analysis Note

Assay of Ni, calc. on dried substance: ≥ 90.0 %
Assay od Al, calc. on dried substance: ≤ 10.0 %
Suitable for hydrogenation catalyst: passes test

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Self-heat. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service