Skip to Content
Merck
All Photos(1)

Key Documents

8.20078

Sigma-Aldrich

4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid monosodium salt

for synthesis

Synonym(s):

4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid monosodium salt, 1-Amino-8-naphthol-3,6-disulfonic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8NNaO7S2
CAS Number:
Molecular Weight:
341.29
MDL number:
UNSPSC Code:
12352107
EC Index Number:
226-736-4
NACRES:
NA.22

Quality Level

form

powder

autoignition temp.

>400 °C

mp

>380 °C

bulk density

550 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)(O)c1cc2c(c(c1)N)c(cc(c2)[S](=O)(=O)O)[O-]

InChI

1S/C10H9NO7S2.Na/c11-8-3-6(19(13,14)15)1-5-2-7(20(16,17)18)4-9(12)10(5)8;/h1-4,12H,11H2,(H,13,14,15)(H,16,17,18);/q;+1/p-1

InChI key

QPILZZVXGUNELN-UHFFFAOYSA-M

Analysis Note

Assay (acidimetric): ≥ 85.0 %
Water (K. F.): 8.0 - 12.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service