Skip to Content
Merck
All Photos(1)

Key Documents

8.18757

Sigma-Aldrich

2,3-Dimercapto-1-propanol

for synthesis

Synonym(s):

2,3-Dimercapto-1-propanol, Dithioglycerine, Dimercaprol, BAL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(SH)CH2SH
CAS Number:
Molecular Weight:
124.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-433-7
NACRES:
NA.22

vapor pressure

7.4 hPa ( 100 °C)

Quality Level

pH

5.0-6.5 (20 °C in H2O, saturated solution)

bp

63-68 °C/0.27 hPa

transition temp

flash point >112 °C

density

1.24 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

SC(CS)CO

InChI

1S/C3H8OS2/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

WQABCVAJNWAXTE-UHFFFAOYSA-N

Application

  • Antibacterial and antifouling properties of lipophilic bismuth compounds: This study discusses the synyhesis of bismuth-2, 3-dimercapto-1-propanol (BAL) in an antibacterial context, highlighting its potential uses in inhibiting biofouling (Badireddy & Chellam, 2014).

Analysis Note

Assay (iodometric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.242 - 1.246
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service