Skip to Content
Merck
All Photos(1)

Key Documents

8.14815

Sigma-Aldrich

Sodium chlorite

(25% solution in water) for synthesis

Synonym(s):

Chlorous acid, sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.22

vapor pressure

20 hPa ( 20 °C)

Quality Level

form

liquid

potency

1136 mg/kg LD50, oral (Rat)

pH

12-13 (20 °C in H2O)

bp

≥100 °C/1013 hPa

mp

<-3 °C

density

1.20-1.25 g/cm3 at 20 °C

storage temp.

2-30°C

Application

Sodium chlorite (NaClO2) can be used as an oxidizing agent for the conversion of:
  • Aldehydes to corresponding carboxylic acids under mild reaction conditions in the presence of scavengers such as sulfamic acid or resorcinol.
  • Primary alcohols to carboxylic acids in the presence of 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) and NaOCl.
  • Aliphatic and aromatic thiols to the corresponding disulfides.
  • Aryl boronic acids to phenols via ipso-hydroxylation.
  • Cyclic benzylic ethers and N-protected cyclic benzylic amines to benzolactones and benzolactams.
  • Alkenes to epoxides using aldehydes as promoters.
  • Alkenes and cycloalkenes to 1, 2 diols via osmium-catalyzed Sharpless asymmetric dihydroxylation.

It can also be used in combination with Br2 to synthesize 2-carboxyazetidin-2-ones from furanylazetidinones.

Analysis Note

Assay (iodometric): 23.0 - 26.0 %
Density (d 20 °C/ 4 °C): 1.200 - 1.220

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 2

Target Organs

spleen

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epoxidation by sodium chlorite with aldehyde-promoted chlorine dioxide formation
Jangam A and Richardson DE
Tetrahedron Letters, 51(50), 6481-6484 (2010)
A simple and effective synthesis of benzolactones and benzolactams by noncatalytic benzylic oxidation of cyclic benzylic ethers and N-protected cyclic benzylic amines with sodium chlorite as an oxidant
Song Ai-Ru, et al.
Synthesis, 44(18), 2903-2909 (2012)
Uses of sodium chlorite and sodium bromate in organic synthesis. A review
Krapcho A Paul
Organic preparations and procedures international, 38(2), 177-216 (2006)
ipso-Hydroxylation of Arylboronic Acids and Boronate Esters by Using Sodium Chlorite as an Oxidant in Water
Gogoi P, et al.
European Journal of Organic Chemistry, 2013(32), 7291-7294 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service