8.02801
ε-Caprolactone
for synthesis
Synonym(s):
ε-Caprolactone, 2-Oxepanone, 6-Hydroxycaproic acid lactone
About This Item
vapor pressure
0.013 hPa ( 20 °C)
Quality Level
Assay
≥98.0% (GC)
form
liquid
autoignition temp.
204 °C
potency
>2000 mg/kg LD50, oral (Rat)
5990 mg/kg LD50, skin (Rabbit)
expl. lim.
1.2-9 % (v/v)
bp
235 °C/1013 hPa
mp
-1.5 °C
transition temp
flash point 127 °C
density
1.07 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O1CCCCC[C]1[O]
InChI
1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2
InChI key
PAPBSGBWRJIAAV-UHFFFAOYSA-N
Application
- Redox Control of Group 4 Metal Ring-Opening Polymerization Activity toward l-Lactide and ε-Caprolactone: Discusses the impact of redox conditions on the efficiency of ring-opening polymerization of ε-caprolactone, important for synthesizing biodegradable plastics (X Wang, JL Brosmer, 2014).
Analysis Note
Density (d 20 °C/ 4 °C): 1.075 - 1.076
Water (K. F.): ≤ 0.10 %
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
228.2 °F - closed cup
Flash Point(C)
109 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service