8.01537
2-Butyne-1,4-diol
(not stabilised) for synthesis
Synonym(s):
2-Butyne-1,4-diol, 1,4-Dimethoxyacetylene
About This Item
vapor pressure
<1 Pa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
solid
autoignition temp.
410 °C
potency
100 mg/kg LD50, oral (Rat)
pH
4-7.5 (23 °C, 100 g/L in H2O)
bp
125-127 °C/3 hPa
mp
54-57 °C
transition temp
flash point 152 °C
solubility
3740 g/L
density
1.11 g/cm3 at 20 °C
bulk density
500 kg/m3
storage temp.
2-30°C
SMILES string
OCC#CCO
InChI
1S/C4H6O2/c5-3-1-2-4-6/h5-6H,3-4H2
InChI key
DLDJFQGPPSQZKI-UHFFFAOYSA-N
Analysis Note
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
305.6 °F - closed cup
Flash Point(C)
152 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service