642511
L-3,3′,5-Triiodothyronine free acid
≥97% (HPLC), solid, respiratory activity stimulator, Calbiochem®
Synonym(s):
L-3,3′,5-Triiodothyronine, Free Acid, High Purity, T₃
About This Item
product name
L-3,3′,5-Triiodothyronine, Free Acid, High Purity, Chromatographically purified.
Quality Level
Assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to off-white
solubility
0.1 M NaOH: 10 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)
InChI key
AUYYCJSJGJYCDS-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Warning
Reconstitution
Other Notes
Williams, G.R., et al. 1994. Endocrinology135, 2375.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1
Target Organs
Thyroid,Cardio-vascular system,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service