Skip to Content
Merck
All Photos(2)

Key Documents

573128

Sigma-Aldrich

STAT3 Inhibitor XIII, C188-9

The Stat3 Inhibitor XIII, C188-9 controls the biological activity of Stat3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

STAT3 Inhibitor XIII, C188-9, N-(1ʹ,2-Dihydroxy-1,2ʹ-binaphthalen-4ʹ-yl)-4-methoxybenzenesulfonamide, F1113-0789

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21NO5S
CAS Number:
Molecular Weight:
471.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to pale beige

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC1=C(C2=C(O)C=CC3=C2C=CC=C3)C=C(NS(C(C=C4)=CC=C4OC)(=O)=O)C5=C1C=CC=C5

InChI key

QDCJDYWGYVPBDO-UHFFFAOYSA-N

General description

A cell-permeable binaphthol-sulfonamide that competes against pY proteins for Stat3 SH2 domain binding. Shown to inhibit G-CSF-induced (100 ng/ml for 15 min following 1 h drug pretreatment) Stat3 tyr705 phosphorylation in 6 human AML cell lines (IC50 from 4.1 to 8.3 µM) and 4 primary pediatric AML cultures (IC50 from 8 to 18 µM). Long-term treatments (24 h for lines and 48 h for primary cultures) result in apoptosis induction (Annexin V-PE staining) in 7 AML lines (EC50 from 8.4 to 43.6 µM) and CD34+ populations from 5 primary pediatric AML samples (EC50 from 0.8 to25 µM). Selectivity studies using Kasumi-1 cells reveals concomitant inhibitions of G-CSF-stimulated TIE2, MATK, JAK1, and HGFR phosphorylation (by 90%, 50%, 40%, and 40%, respectively; 10 µM C188-9), but not 15 other phosphoproteins (≤30% inhibition), including pERK1/2 and pAKT.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Redell, M.S., et al. 2011. Blood112, 355.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liuqing Ge et al.
Immunity, inflammation and disease, 12(2), e1160-e1160 (2024-02-28)
Regulatory T cells (Tregs) play an important role in inflammatory bowel diseases (IBDs) through modulating intestinal inflammation. However, the factors affecting Treg function and plasticity during IBD progression are not thoroughly disclosed. The current study aims to reveal new molecular
Kimberly M Arnold et al.
International journal of radiation biology, 96(4), 434-447 (2019-12-19)
Purpose: Pro-inflammatory cytokines within the tumor microenvironment, such as IL-6, contribute to the maintenance of stem cells and promote their survival following treatment. The IL-6/STAT3 pathway is a key regulator of genes involved in cancer progression. Activation of STAT3 promotes
Shanchun Guo et al.
Cellular and molecular life sciences : CMLS, 80(9), 270-270 (2023-08-29)
We previously reported that TRPM7 regulates glioma cells' stemness through STAT3. In addition, we demonstrated that FOSL1 is a response gene for TRPM7, and the FOSL1 gene serves as an oncogene to promote glioma proliferation and invasion. In the present
Li Wang et al.
Disease models & mechanisms, 14(11) (2021-09-21)
Vascular permeability triggered by inflammation or ischemia promotes edema, exacerbates disease progression and impairs tissue recovery. Vascular endothelial growth factor (VEGF) is a potent inducer of vascular permeability. VEGF plays an integral role in regulating vascular barrier function physiologically and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service