Skip to Content
Merck
All Photos(1)

Key Documents

540211

Sigma-Aldrich

PTP Inhibitor IV

The PTP Inhibitor IV, also referenced under CAS 329317-98-8, controls the biological activity of PTP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PTP Inhibitor IV, bis(4-Trifluoromethylsulfonamidophenyl)-1,4-diisopropylbenzene, Protein Tyrosine Phosphatase Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26F6N2O4S2
CAS Number:
Molecular Weight:
608.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 10 mg/mL
acetonitrile: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)[S](=O)(=O)Nc1ccc(cc1)C(C)(C)c2ccc(cc2)C(C)(C)c3ccc(cc3)N[S](=O)(=O)C(F)(F)F

InChI

1S/C26H26F6N2O4S2/c1-23(2,19-9-13-21(14-10-19)33-39(35,36)25(27,28)29)17-5-7-18(8-6-17)24(3,4)20-11-15-22(16-12-20)34-40(37,38)26(30,31)32/h5-16,33-34H,1-4H3

InChI key

JWNAVYMNAQDLHX-UHFFFAOYSA-N

General description

An uncharged, 1,4-di-substituted, phenyl-linked bis-trifluoromethylsulfonamido (TFMS) phosphate mimetic that acts as a potent, reversible, competitive, and active-site directed inhibitor of protein tyrosine phosphatases (PTP; IC50 = 1.8, 2.5, 8.4, 13, 20, 6.4, and 6.7 µM for SHP-2, PTP1B, PTP-ε, PTP-Meg-2, PTP-ς, PTP-β, and PTP-µ, respectively).
An uncharged, 1,4-di-substituted, phenyl-linked, bis-trifluoromethylsulfonamido (TFMS) phosphate mimetic that acts as a potent, reversible, substrate competitive, active-site-directed inhibitor of protein tyrosine phosphatases (PTP). Reported to inhibit SHP-2 (IC50 = 1.8 µM), PTP1B (IC50 = 2.5 µM), PTP-ε (IC50 = 8.4 µM), PTP-Meg-2 (IC50 = 13 µM), PTP-ς (IC50 = 20 µM), PTP-β (IC50 = 6.4 µM), and PTP-µ (IC50 = 6.7 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
SHP-2
Product competes with ATP.
Reversible: yes
Target IC50: 1.8, 2.5, 8.4, 13, 20, 6.4, and 6.7 µM for SHP-2, PTP1B, PTP-ε, PTP-Meg-2, PTP-σ, PTP-β, and PTP-µ, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Huang, P., et al. 2003. Bioorg. Med. Chem.11, 1835.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Xu et al.
Annals of translational medicine, 9(4), 350-350 (2021-03-13)
Ischemia reperfusion (IR)-induced acute kidney injury (AKI) is accompanied by increased inflammatory response and oxidative stress. Eriocitrin is a flavonoid that is mainly derived from lemon or citrate juice. It exhibits various pharmacological effects and is known to have antioxidant
D G Semaan et al.
Journal of ethnopharmacology, 203, 39-46 (2017-03-28)
Ethno-botanical information from diabetic patients in Cuba led to the identification of Allophylus cominia as a possible source of new drugs for the treatment of type 2 diabetes mellitus (T2-DM). Chemical characterization of the extracts from A. cominia was carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service