Skip to Content
Merck
All Photos(1)

Key Documents

5.05732

Sigma-Aldrich

GBR 12909 Dihydrochloride

Synonym(s):

GBR 12909 Dihydrochloride, 1-(2-Bis(4-fluorophenyl)methoxy)ethyl)-4-(3-phenylpropyl)piperazine Hydrochloride, Dopamine Active Transporter Inhibitor, GBR 12909, DAT Inhibitor, GBR 12909, GBR12909

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32F2N2O · 2HCl
CAS Number:
Molecular Weight:
523.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

1 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 100 mM

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(OCCN3CCN(CC3)CCCc4ccccc4)c2ccc(cc2)F.[Cl-].[Cl-].[H+].[H+]

InChI

1S/C28H32F2N2O.2ClH/c29-26-12-8-24(9-13-26)28(25-10-14-27(30)15-11-25)33-22-21-32-19-17-31(18-20-32)16-4-7-23-5-2-1-3-6-23;;/h1-3,5-6,8-15,28H,4,7,16-22H2;2*1H

InChI key

MIBSKSYCRFWIRU-UHFFFAOYSA-N

General description

A potent inhibitor of dopamine active transporter (DAT, Ki = 1 nM, IC50 = 40 and 51 nM) with >100 fold selectivity over noradrenalin and 5-HT re-uptake transporter. Shown to produce behavioral profile in mice for bipolar mania disease model and ADHD disease models.

Biochem/physiol Actions

Primary Target
DAT

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Young, J. et al. 2010. Psychopharm.208, 443.

Hewitt, K., et al. 2009. Neuropharm.57, 678.
Prisinzano, T. et al. 2002. J. Med. Chem.45, 4371.
Andersen, P., et al. 1989. Eur. J. Pharmacol.166, 493.
Heikkila, R., et al. 1984. Eur. J. Pharmacol.103, 142.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service