Skip to Content
Merck
All Photos(1)

Key Documents

401480

Sigma-Aldrich

IKK Inhibitor III, BMS-345541

IKK Inhibitor III, BMS-345541, CAS 445430-58-0, is a cell-permeable, potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC₅₀ = ~ 300 nM).

Synonym(s):

IKK Inhibitor III, BMS-345541, 4-(2ʹ-Aminoethyl)amino-1,8-dimethylimidazo[1,2-a]quinoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N5 · 2C2HF3O2
CAS Number:
Molecular Weight:
483.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥96% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21c(ncc2C)c(nc3c1cc(cc3)C)NCCN

InChI

1S/C14H17N5/c1-9-3-4-11-12(7-9)19-10(2)8-17-14(19)13(18-11)16-6-5-15/h3-4,7-8H,5-6,15H2,1-2H3,(H,16,18)

InChI key

PSPFQEBFYXJZEV-UHFFFAOYSA-N

General description

A cell-permeable quinoxaline compound that displays anti-inflammatory properties. Acts as a potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC50 = ~ 300 nM). Exhibits ~10-fold greater selectivity over IKK-1 (IC50 = ~ 4 µM) and no activity towards IKKε and a panel of more than 15 unrelated protein kinases even at concentrations as high as 100 µM. Inhibits cellular IκBα phosphorylation (IC50 = 4 µM in THP-1 cells) and LPS-induced cytokine production both in vitro (IC50 = 1-5 µM in THP-1 cells) and in vivo (IC50 = 10 mg/kg in mice). Effectively blocks inflammation and joint destruction in a murine arthritis model.
A cell-permeable, potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC50 ~300 nM) that displays anti-inflammatory properties. Exhibits ~10-fold greater selectivity for IKK-2 over IKK-1 (IC50 ~4.0 µM) and does not inhibit IKKε and a panel of more than 15 unrelated protein kinases even at concentrations as high as 100 µM. Inhibits cellular IκBα phosphorylation (IC50 = 4 µM in THP-1 cells) and LPS-induced cytokine production both in vitro (IC50 = 1-5 µM in THP-1 cells) and in vivo (IC50 = 10 mg/kg in mice). Effectively blocks inflammation and joint destruction in a murine arthritis model.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: ~300 nM against IKK-2; 4 µM against cellular IκBα phosphorylation in THP-1 cells; 1-5 µM LPS-induced cytokine production in THP-1 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as bis-TFA salt.

Other Notes

Townsend, R.M., et al. 2004. Transplantation77, 1090.
MacMaster, J.F., et al. 2003. Inflamm. Res.52, 508.
McIntyre, K.W., et al. 2003. Arthritis Rheum.48, 2652.
Burke, J. R., et al. 2003. J. Biol. Chem.278, 1450.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reshma Raghava Kurup et al.
Scientific reports, 12(1), 13362-13362 (2022-08-04)
The RNA binding protein ADAR3 is expressed exclusively in the brain and reported to have elevated expression in tumors of patients suffering from glioblastoma compared to adjacent brain tissue. Yet, other studies have indicated that glioblastoma tumors exhibit hemizygous deletions
Weili Wang et al.
Nature communications, 13(1), 4495-4495 (2022-08-03)
While hypoxia promotes carcinogenesis, tumour aggressiveness, metastasis, and resistance to oncological treatments, the impacts of hyperoxia on tumours are rarely explored because providing a long-lasting oxygen supply in vivo is a major challenge. Herein, we construct micro oxygen factories, namely
Owen S Skinner et al.
Nature metabolism, 5(5), 765-776 (2023-05-18)
Glucose is vital for life, serving as both a source of energy and carbon building block for growth. When glucose is limiting, alternative nutrients must be harnessed. To identify mechanisms by which cells can tolerate complete loss of glucose, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service