Skip to Content
Merck
All Photos(1)

Key Documents

400064

Sigma-Aldrich

Anti-Glucose Transporter-4 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Glut-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A4(6517)

General description

Anti-Glucose Transporter-4, rabbit polyclonal, recognizes the ~40-43 kDa GLUT-4. It is validated for use in ELISA, Western blotting, immunoprecipitation, and immunohistochemistry on frozen sections.
Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~40-43 kDa Glut-4 protein.
Recognizes the ~40-43 kDa GLUT-4 protein.

Immunogen

Mouse
a synthetic peptide corresponding to amino acids near the C-terminus of mouse GLUT-4, conjugated to KLH

Application

ELISA (0.5-1 µg/ml)

Frozen Sections (2-20 µg/ml)

Immunoblotting (1-10 µg/ml; see comments)

Immunoprecipitation (1-10 µg/100 µg tissue membrane)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 0.1% BSA, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Does not cross-react with Hep G2-type transporter or other Glut isoforms. Important: Do not heat samples prior to immunoblotting, as heating will result in protein aggregation. Variables associated with assay conditions will dictate the optimal working dilution.
Mueckler, M. 1994. Eur. J. Biochem. 219, 713.
James, D.E., et al. 1989. Nature 338, 83.
James, D.E., et al. 1989. Proc. Natl. Acad. Sci. USA 86, 8368.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae-Heung An et al.
Journal of exercise rehabilitation, 12(3), 163-170 (2016-07-16)
The purpose of this study was to investigate the expression of lipogenesis- and lipolysis-related genes and proteins in skeletal muscles after 12 weeks of resistance training. Sprague-Dawley rats (n=12) were randomly divided into control (resting) and resistance training groups. A
Xue-Han Ning et al.
American journal of physiology. Heart and circulatory physiology, 292(1), H333-H341 (2006-09-05)
Hypoxia-inducible factor 1alpha (HIF-1alpha) transcriptionally activates multiple genes, which regulate metabolic cardioprotective and cross-adaptive mechanisms. Hypoxia and several other stimuli induce the HIF-1alpha signaling cascade, although little data exist regarding the stress threshold for activation in heart. We tested the
Malcolm Grohmann et al.
The Journal of physiology, 568(Pt 1), 229-242 (2005-08-06)
We have developed a primary skeletal muscle cell culture model derived from normal prepubertal children to investigate the effects of insulin-like growth factor-I (IGF-I), insulin-like growth factor binding protein-3 (IGFBP-3) and tumour necrosis factor alpha (TNFalpha) on growth, differentiation and
Young Ju Lee et al.
Molecular medicine reports, 7(5), 1571-1578 (2013-04-03)
Alzheimer's disease (AD) is closely associated with significant defects in glucose metabolism. To investigate whether AD pathology induced by overexpression of human mutant presenilin 2 (PS2) protein induces changes in glucose metabolism, glucose‑related factors were analyzed in the brain of
Eric B Taylor et al.
The Journal of biological chemistry, 283(15), 9787-9796 (2008-02-16)
The Akt substrate of 160 kDa (AS160) is phosphorylated on Akt substrate (PAS) motifs in response to insulin and contraction in skeletal muscle, regulating glucose uptake. Here we discovered a dissociation between AS160 protein expression and apparent AS160 PAS phosphorylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service