Skip to Content
Merck
All Photos(1)

Documents

34860-M

Sigma-Aldrich

Methanol

≥99.9%, suitable for HPLC

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

product name

Methanol, suitable for HPLC, ≥99.9%

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm
<0.03% water (Karl Fischer)

evapn. residue

<0.0005%

color

APHA: ≤10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

λ

neat

UV absorption

λ: 205 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.60
λ: 220 nm Amax: ≤0.30
λ: 230 nm Amax: ≤0.20
λ: 235 nm Amax: ≤0.10
λ: 240 nm Amax: ≤0.10
λ: 260 nm Amax: ≤0.04
λ: 280 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.01

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol is an organic solvent that can be synthesized from syngas in the presence of CuO/ZnO/Al2O3 catalysts. It is an ideal candidate as a hydrogen source in fuel cell technology due to its high H/C ratio, low propensity for soot generation, relatively low reforming temperature and as it exists in liquid state at room temperature. In a direct methanol fuel cell (DMFC), methanol undergoes oxidation with air to generate electricity. The olefins (ethylene or propylene) formed from methanol via MTO (methanol-to-olefins) process, can be an alternate to oil and gas to produce hydrocarbon fuels.
Thermochemical conversion of methanol to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.

Application

Methanol (MeOH) has been used in the wholemount immunofluorescence studies of embryonic tissues.
It may be used in the following studies:
  • Colony forming unit-fibroblast assay of bone marrow mononuclear cells.
  • As solvent for the preparation of extracts of hyphae of Aspergillus for the estimation of gliotoxin by reversed phase-HPLC.
  • Immunofluorescence studies.
  • To compose the eluent for the ion-pair reverse-phase HPLC isolation of nucleotides and their decomposition products.
Methanol can be used as a mobile phase in reversed-phase liquid chromatography.

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Important notice
This product will be phased out towards early 2019. There is a replacement product available : 34860

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Review of methanol reforming-Cu-based catalysts, surface reaction mechanisms, and reaction schemes.
Yong ST, et al.
International Journal of Hydrogen Energy, 38(22), 9541-9552 (2013)
R A Cunha et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(6), 1987-1995 (1998-05-15)
ATP analogs substituted in the gamma-phosphorus (ATPgammaS, beta, gamma-imido-ATP, and beta,gamma-methylene-ATP) were used to probe the involvement of P2 receptors in the modulation of synaptic transmission in the hippocampus, because their extracellular catabolism was virtually not detected in CA1 slices.
Jonas Ahnfelt-Rønne et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 55(9), 925-930 (2007-05-05)
We have developed a wholemount immunofluorescence protocol for the simultaneous detection of up to three proteins in mouse and chicken embryos. Combined with Murray's clearing reagent (BABB) and microscope objectives with long working ranges and high numerical apertures mounted on
Emer P Reeves et al.
Mycopathologia, 158(1), 73-79 (2004-10-19)
Aspergillus fumigatus is a pathogenic fungus capable of causing both allergic lung disease and invasive aspergillosis, a serious, life-threatening condition in neutropenic patients. Aspergilli express an array of mycotoxins and enzymes which may facilitate fungal colonisation of host tissue. In
Robert E Schwartz et al.
The Journal of clinical investigation, 109(10), 1291-1302 (2002-05-22)
We have derived from normal human, mouse, and rat postnatal bone marrow primitive, multipotent adult progenitor cells (MAPCs) that can differentiate into most mesodermal cells and neuroectodermal cells in vitro and into all embryonic lineages in vivo. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service