233165
Clioquinol
A metal ion chelator that crosses the blood brain barrier and acts as a neurotoxic antibiotic.
Synonym(s):
Clioquinol, 5-Chloro-7-iodo-8-hydroxyquinoline
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
solubility
ethanol: 1 mg/mL
methanol: 1.5 mg/mL
DMSO: 20 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
Ic1c(c2ncccc2c(c1)Cl)O
InChI
1S/C9H5ClINO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H
InChI key
QCDFBFJGMNKBDO-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Metal ion chelator
Packaging
Warning
Reconstitution
Other Notes
Regland, B., et al. 2001. Dement. Geriatr. Cogn. Disord.12, 408.
Helmuth, L., et al. 2000. Science290, 1273.
Tateishi, J., et al. 2000. Neuropathology20, Suppl: S20-4.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service