Skip to Content
Merck
All Photos(1)

Key Documents

219477

Sigma-Aldrich

Cdk4 Inhibitor II, NSC 625987

The Cdk4 Inhibitor II, NSC 625987, also referenced under CAS 141992-47-4, controls the biological activity of Cdk4. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk4 Inhibitor II, NSC 625987, 1,4-Dimethoxyacridine-9(10H)-thione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO2S
CAS Number:
Molecular Weight:
271.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

ethanol: 2.5 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S=[c]1c2c([nH]c3c1cccc3)c(ccc2OC)OC

InChI

1S/C15H13NO2S/c1-17-11-7-8-12(18-2)14-13(11)15(19)9-5-3-4-6-10(9)16-14/h3-8H,1-2H3,(H,16,19)

InChI key

KFAKESMKRPNZTM-UHFFFAOYSA-N

General description

A potent, reversible, and substrate-competitive inhibitor of Cdk4/cyclin D1 (IC50 = 200 nM). Displays ~500-fold greater selectivity for Cdk4/cyclin D1 over Cdc2/cyclin A (Cdk1/cyclin A), Cdk2/cyclin A, and Cdk2/cyclin E (IC50 >100 µM).
A thioacridone compound that acts as a reversible and potent substrate-competitive inhibitor of Cdk4/cyclin D1 (IC50 = 200 nM). Displays ~500-fold greater selectivity for Cdk4/cyclin D1 over Cdc2/cyclin A (Cdk1/cyclin A), Cdk2/cyclin A, and Cdk2/cyclin E (IC50 >100 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
CDK4/D1
Product does not compete with ATP.
Reversible: yes
Target IC50: 200 nM against CDK4/D1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kubo, A., et al. 1999. Clin. Cancer Res.5, 4279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service