Skip to Content
Merck
All Photos(1)

Key Documents

200487

Sigma-Aldrich

β-Amyloid Oligomer Inhibitor, K01-162

The β-Amyloid Oligomer Inhibitor, K01-162 controls the biological activity of β-Amyloid Oligomer.

Synonym(s):

β-Amyloid Oligomer Inhibitor, K01-162, AβO Inhibitor, K01-162

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14BrN
Molecular Weight:
288.18
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

color

white to off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell permeable, fluorene compound that destabilizes the aggregation state of β-amyloid oligomers (AβO) by directly targeting AβO and inhibiting AβO binding to synapses. It displays full MC65 protection at 125 nM, with an EC50 of 80 nM in a neuronal MC65 culture-based assay, and substantially reduces intraneuronal AβO in primary neuronal culture at concentrations between 100 nM and 10 µM. Furthermore, it is shown to significantly reduce amyloid levels by 50% compared with control in vivo in a 5xFAD mice cerebral Aβ amyloidosis model when dosed intracerebroventricularly without apparent toxicity.
A cell permeable, fluorene compound that destabilizes the aggregation state of β-amyloid oligomers (AβO) by directly targeting AβO and inhibiting AβO binding to synapses. It displays full MC65 protection at 125 nM, with an EC50 of 80 nM in a neuronal MC65 culture-based assay, and substantially reduces intraneuronal AβO in primary neuronal culture at concentrations between 100 nM and 10 µM. Furthermore, it is shown to significantly reduce amyloid levels by 50% compared with control in vivo in a 5xFAD mice cerebral Aβ amyloidosis model when dosed intracerebroventricularly without apparent toxicity.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Hong, H-S., et al. 2010. Neurobiol. Aging31, 1690.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dusan Mrdenovic et al.
ACS chemical neuroscience, 12(3), 531-541 (2021-01-23)
Alzheimer's disease (AD) is characterized by progressive neurodegeneration associated with amyloid β (Aβ) peptide aggregation. The aggregation of Aβ monomers (AβMs) leads to the formation of Aβ oligomers (AβOs), the neurotoxic Aβ form, capable of permeating the cell membrane. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service