All Photos(1)
About This Item
UNSPSC Code:
12352302
eCl@ss:
32160405
NACRES:
NA.41
grade:
for analytical purposes
form:
solid
Recommended Products
grade
for analytical purposes
Quality Level
form
solid
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
shipped in
wet ice
Application
Potent and selective inhibitor of PKA.
Biochem/physiol Actions
Inhibitor Type: Kinase
Physical form
lyophilized powder
Storage and Stability
2 years at -20°C in a desiccation chamber.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
F J Johannes et al.
European journal of biochemistry, 227(1-2), 303-307 (1995-01-15)
In order to investigate regulatory mechanisms and to identify potential substrates of a novel member of the protein kinase C (PKC) family, PKC mu, specific antibodies have been raised against unique amino- and carboxy-terminal regions. PKC mu kinase activity was
T Chijiwa et al.
The Journal of biological chemistry, 265(9), 5267-5272 (1990-03-25)
A newly synthesized isoquinolinesulfonamide, H-89 (N-[2-(p-bromocinnamylamino)ethyl]-5-isoquinoline-sulfonamide), was shown to have a potent and selective inhibitory action against cyclic AMP-dependent protein kinase (protein kinase A), with an inhibition constant of 0.048 +/- 0.008 microM. H-89 exhibited weak inhibitory action against other
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service