182540-M
Aromatase Inhibitor I
The Aromatase Inhibitor I, also referenced under CAS 331684-05-0, controls the biological activity of Aromatase.
Synonym(s):
Aromatase Inhibitor I, 4-(Imidazolylmethyl)-1-nitro-9H-9-xanthenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H11N3O4
CAS Number:
Molecular Weight:
321.29
UNSPSC Code:
12352200
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 25 mg/mL
storage temp.
−20°C
General description
A potent, competitive, nonsteroidal inhibitor of aromatase (P450arom; IC50 = 40 nM for human aromatase). Reported to be more potent than fadrozole in inhibiting aromatase activity. Does not significantly inhibit 17α-hydroxylase.
A potent, selective, nonsteroidal, competitive inhibitor of aromatase (P450arom) (IC50 = 40 nM for human aromatase) Reported to be more potent than fadrozole in inhibiting aromatase activity. Does not significantly inhibit human 17α-hydroxylase (P450 17) (4% inhibition at 2.5 µM).
Biochem/physiol Actions
Primary Target
P450 arom.
P450 arom.
Target IC50: 40 nM against P450arom
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Recanatini, M., et al. 2001. J. Med. Chem.44, 672.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service