Key Documents
1.52614
Supelco
Electronic Unit
greener alternative
Replacement part
About This Item
UNSPSC Code:
39121400
NACRES:
NB.21
Quality Level
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
storage temp.
2-30°C
General description
The Electronic unit is a spare part suitable for the TLC Explorer Documentation System (Cat. No. 1.52610.0001 ). The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates. It offers a safe, fast and flexible way to visually inspect and digitally record images of TLC plates after their chromatographic development workflow in the laboratory. For detecting different substance classes, the device contains three different LED light sources (white light (VIS), UVA (366 nm) and UVC (254 nm), which illuminate the plates from above in direct-light mode. Recorded plate images can be evaluated to generate reports or export the data for further processing with other systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Visit the Document Library
Related Content
Discover an efficient and automated software for accurate and reliable qualitative and quantitative TLC analyses. Enjoy a hassle-free plug-and-play experience and easily connect through WiFi or LAN for seamless connectivity.
Discover an efficient and automated software for accurate and reliable qualitative and quantitative TLC analyses. Enjoy a hassle-free plug-and-play experience and easily connect through WiFi or LAN for seamless connectivity.
Discover an efficient and automated software for accurate and reliable qualitative and quantitative TLC analyses. Enjoy a hassle-free plug-and-play experience and easily connect through WiFi or LAN for seamless connectivity.
Qualitative and quantitative TLC analyses. Automated software for hassle-free, quick, and precise results. Plug-and-play setup with intuitive software.
See All