1.11037
tri-Sodium citrate
anhydrous, EMPROVE® ESSENTIAL, USP, FCC, E 331
Pharma Manufacturing
Synonym(s):
tri-Sodium citrate
About This Item
Agency
USP
Quality Level
reg. compliance
FCC
product line
EMPROVE® ESSENTIAL
form
solid
pH
7.5-9.0 (20 °C, 10 g/L in H2O)
mp
150 °C (decomposition)
solubility
425 g/L
density
1.76 g/cm3 at 18 °C
application(s)
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
solid formulation
storage temp.
2-30°C
SMILES string
[Na+].[Na+].[Na+].[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-]
InChI
1S/C6H8O7.3Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3
InChI key
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
also commonly purchased with this product
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service