Skip to Content
Merck
All Photos(1)

Key Documents

1.10958

Supelco

Kjeldahl tablets

for Wieninger method 5g/tablet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352402
NACRES:
NB.21

Quality Level

form

tablet

specific analyte(s)

nitrogen

technique(s)

photometry: suitable

pH

5 (20 °C, 200 g/L in H2O)

solubility

200 g/L

storage temp.

no temp limit

General description

Type: Wieninger catalyst Composition:
Na2SO4 96.5%
CuSO4 1.5%
Se 2.0%

Analysis Note

Appearance: grey tablets
Application test: conforms
Grey colour of the product is caused by raw material and does not affect the application.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.

Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.

Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.

Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service