Skip to Content
Merck
All Photos(1)

Key Documents

110175-M

Millipore

Acetyl-Pepstatin

An aspartyl protease inhibitor that acts as an effective inhibitor of HIV-1 proteinase (Ki = 20 nM at pH 4.7) and HIV-2 proteinase (Ki = 5 nM at pH 4.7).

Synonym(s):

Acetyl-Pepstatin, Ac-Val-Val-(3S,4S)-Sta-Ala-(3S,4S)-Sta-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H57N5O9
CAS Number:
Molecular Weight:
643.81
MDL number:
UNSPSC Code:
12352202

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

50% acetic acid: 5 mg/mL

storage temp.

−20°C

SMILES string

N([C@H]([C@@H](O)CC(=O)N[C@@H](C)C(=O)N[C@H]([C@@H](O)CC(=O)O)CC(C)C)CC(C)C)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)C(C)C)C(C)C

InChI

1S/C31H57N5O9/c1-15(2)11-21(35-30(44)28(18(7)8)36-31(45)27(17(5)6)33-20(10)37)23(38)13-25(40)32-19(9)29(43)34-22(12-16(3)4)24(39)14-26(41)42/h15-19,21-24,27-28,38-39H,11-14H2,1-10H3,(H,32,40)(H,33,37)(H,34,43)(H,35,44)(H,36,45)(H,41,42)/t19-,21-,22-,23-,24-,27-,28-/m0/s1

InChI key

WKYBEGDEGRCZNF-LBTYKNIQSA-N

General description

An aspartyl protease inhibitor that acts as an effective inhibitor of HIV-1 proteinase (Ki = 20 nM at pH 4.7) and HIV-2 proteinase (Ki = 5 nM at pH 4.7). Acts as a substrate-analog inhibitor.

Biochem/physiol Actions

Primary Target
HIV-1 proteinase
Target Ki: 20 nM at pH 4.7 against HIV-1 proteinase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Ulrich, B., et al. 2000. Protein Sci.9, 2232.
Song, K., et al. 1992. Kidney Int.42, 639.
Richards, A.D., et al. 1989. FEBS Lett. 247, 113.
Richards, A.D., et al. 1989. FEBS Lett. 253, 214.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service