Skip to Content
Merck
All Photos(1)

Key Documents

1.09889

Supelco

Buffer concentrate

(boric acid/potassium chloride/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 9.00 ± 0.02 (20°C) Titrisol®

Synonym(s):

Buffer concentrate for 500 ml buffer solution, pH 9.00 ± 0.02 (20°C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161705
NACRES:
NA.25

grade

analytical standard

Quality Level

Agency

according to DIN 19266

product line

Titrisol®

pH

8.9 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The Titrisol® reference buffer concentrate [boric acid/potassium chloride/sodium hydroxide; pH=9.00±0.02 (20 °C)] is analyzed in the ISO/IEC 17025 accredited calibration laboratory with a combined glass electrode and 5-point calibration according to DIN 19268 against a certified primary standard reference material following DIN 19266. The Titrisol® concentrated buffer material combines ease of use and maximum precision for the routine calibration and monitoring of pH instruments. To prepare the ready-to-use pH buffer solution, simply dilute the Titrisol® buffer concentrate.

Application

  • Buffer concentrate in viral nucleic acid preamplification: A study utilized buffer concentrate in various viral nucleic acid preamplification pipelines for sewage viral metagenomics, enhancing the accuracy and efficiency of viral detection methods (Fernandez-Cassi and Kohn, 2024).
  • Buffer concentrate in myocardial slice cultures: Research on rabbit myocardial slices demonstrated the critical role of buffer concentrate in maintaining tissue stability and function under various experimental conditions, crucial for physiological studies (Baron et al., 2024).
  • Buffer concentrate in fluorescence spectroscopy: A study explored the interaction of erythrosine B with cardiac drugs, using buffer concentrate to stabilize reaction conditions in fluorescence, scattering, and absorbance spectroscopy (AlSalem et al., 2024).
  • Buffer concentrate in solid-state NMR: The quantification of residual water in pharmaceutical solutions was improved using buffer concentrate in solid-state NMR, highlighting its importance in ensuring the quality and stability of pharmaceutical formulations (Du and Su, 2024).
  • Buffer concentrate in electrochemical detection: Buffer concentrate facilitated the electrochemical detection of iodide in nuclear emergency tablets, proving essential for precise and reliable analytical applications in emergency response scenarios (Alqibthiyah et al., 2024).

Features and Benefits

  • Traceable to primary standard reference materials from NIST and PTB
  • Analytical standard Grade
  • Uncertainty data and traceability are documented in a comprehensive and batch-specific Certificate of Analysis.

Analysis Note

pH-value (20 °C): 9.00 +/- 0.02
Measured after dilution to 500 ml with a combination glass electrode after multiple point calibration (according to DIN 19268) with reference materials for the preparation of standard buffer solutions according to DIN 19266.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

standard

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service