Skip to Content
Merck
All Photos(1)

Key Documents

1.08603

Millipore

Triton X-100

for analysis

Synonym(s):

Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

potency

1900-5000 mg/kg LD50, oral (Rat)
>3000 mg/kg LD50, skin (Rabbit)

mol wt

micellar avg mol wt 80,000
average mol wt 625

aggregation number

100-155

pH

5.0-8.0 (20 °C, 10 g/L in H2O)

CMC

0.2-0.9 mM (20-25°C)

bp

>200 °C/1013 hPa

transition temp

flash point 251 °C
cloud point 65 °C
pour point ~7 °C

density

1.07 g/cm3 at 20 °C

HLB

13.5

storage temp.

15-25°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • A liposome encapsulated methylene blue-mediated electrochemical and UV-visible dual mode split-type immunoassay for the detection of 17 beta-estradiol.: This study presents a dual detection method incorporating Triton X-100 to enhance the performance of assays in detecting hormonal levels critical for biochemistry applications (Xia et al., 2024).

  • From Antagonism to Enhancement: Triton X-100 Surfactant Affects Phenanthrene Interfacial Biodegradation by Mycobacteria through a Shift in Uptake Mechanisms.: This research explores how Triton X-100 modifies biodegradation processes at the microbial interface, pivotal for biochemists studying environmental and biochemical applications of surfactants (Wang et al., 2024).

  • Tumors cells with mismatch repair deficiency induce hyperactivation of pyroptosis resistant to cell membrane damage but are more sensitive to co-treatment of IFN-gamma and TNF-alpha to PANoptosis.: Investigates the complex interactions between cellular processes influenced by Triton X-100, which are crucial for understanding the surfactant′s role in biochemistry and cancer research (Li et al., 2024).

  • Subcranial Encephalic Temnograph-Shaped Helmet for Brain Stroke Monitoring.: This study utilizes Triton X-100 to create phantom brain tissues to investigate medical monitoring systems for brain stroke (Cuccaro et al., 2024).

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Linkage

Replaces: 8603-30; 8603

Analysis Note

Identity (IR-spectrum): passes test
pH-value (10 g/l): 6.0 - 8.0
Density (d 20/4): 1.064 - 1.067
Cloud point: 63 - 69 °C
Ethylene oxide (GC): ≤ 30 ppm
Dioxane (GC): ≤ 20 ppm
Sulfated ash (600 °C): ≤ 0.2
Water (according to Karl Fischer): ≤ 0.2

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service