Skip to Content
Merck
All Photos(2)

Key Documents

1.05593

Supelco

Sodium hydroxide solution

21%, for analysis, EMSURE®

Synonym(s):

Sodium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Sodium hydroxide solution 21%, for analysis EMSURE®

vapor pressure

3 mmHg ( 37 °C)

Quality Level

product line

EMSURE®

form

liquid

concentration

19.0-21.8% NaOH

impurities

≤0.001% Fe (Iron)
≤0.002% Heavy metals (as Pb)
≤0.005% Chloride (Cl)
≤0.005% Sulfate (SO4)
≤0.2% Carbonate (as Na2CO3)

pH

>11 (20 °C in H2O)

density

1.219 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide solution 21% (EMSURE® grade product) is optimized for regulated analyses and highly demanding lab applications. It also offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. This reagent is ideal for demanding analytical procedures that need low detection limits and maximum sensitivity.

Application


  • Recycling of Geopolymer Concrete: Discusses the utilization of sodium hydroxide solution in the recycling processes of metakaolin/slag-based geopolymer concrete, enhancing sustainability in construction materials under freeze-thaw conditions (Liu M, Liu H, et al., 2024).

  • Recovery of Thorium and Tungsten: Details the use of sodium hydroxide solution in hybrid electrolysis-nanofiltration processes for the recovery of valuable metals from industrial waste, contributing to environmental sustainability and resource recovery (Man GT, Albu PC, et al., 2024).

  • CO2 Adsorption Dynamics: Highlights the use of sodium hydroxide solution impregnated in activated carbon for CO2 adsorption, showcasing its effectiveness in environmental pollution control and providing insights into optimizing adsorption conditions using response surface methodology (Borisut P, Tangsathitkulchai C, et al., 2024).

  • Quantitation of Hydroxycinnamic Acid: Describes a new HPLC-UV method utilizing hydrolyzation with sodium hydroxide solution for the quantitation of trans-p-hydroxycinnamic acid and its esters in plant leaves, advancing analytical chemistry in pharmaceutical and nutraceutical research (Lu SH, Liang XN, et al., 2023).

Features and Benefits

  • Most extensive specifications worldwide: Tested for up to 70 parameters, extraordinary purity, very low limiting values
  • Greater accuracy and control of analyses      
  • Optimized for highly critical and demanding analyses      
  • Ideal for method development      
  • No interference or contamination due to unknown impurities

Analysis Note

Assay (acidimetric): 19.0 - 21.8 %
Carbonate (as Na₂CO₃): ≤ 0.2 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service