1.01775
Petroleum benzine
for analysis boiling range 40-60°C EMSURE® ACS,ISO
Synonym(s):
Petroleum ether
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. ISO
vapor pressure
370 hPa ( 25 °C)
product line
EMSURE®
form
liquid
autoignition temp.
260 °C (DIN 51794)
potency
>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
expl. lim.
1.0-7.4 % (v/v)
impurities
≤0.0003 meq/g Acidity
≤0.005% Sulfur compounds (as S)
≤0.02% Aromatics (as benzene)
evapn. residue
≤0.001%
color
APHA: ≤10
kinematic viscosity
0.45 cSt(20 °C)
bp
36-83 °C/1013 hPa
mp
<-80 °C
iodine value
≤0.3
peroxide value
≤0.8
transition temp
flash point <-21 °C
density
0.645-0.665 g/cm3 at 15 °C
cation traces
Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Ginkgo Biloba L. Exocarp Extracts: Utilizing petroleum benzine for solvent extraction, this research explores the anti-microbial potential of Ginkgo biloba L. exocarp extracts against methicillin-resistant Staphylococcus aureus, indicating its significant role in pharmaceutical applications (Wei et al., 2024).
- Creating Mesopores in ZSM-5 Catalyst for Aromatics Production: This application of petroleum benzine in the synthesis of biodiesel-derived waste glycerol showcases innovative use in catalytic processes to enhance aromatic production, beneficial for both environmental and industrial chemistry (Kumar et al., 2024).
- High-Speed Countercurrent Chromatography for Bioactive Neolignan Isomers: Demonstrating the use of petroleum benzine in chromatographic techniques, this method significantly aids in the isolation of complex organic compounds, crucial for advanced research in natural product chemistry (Xiao et al., 2024).
- Hydrocarbon Ingestions and Poison Control: While indirectly related, the study uses data on hydrocarbon ingestions to emphasize the importance of safety and environmental impact assessments in the use of petroleum products, including benzine, within public health contexts (Palmer et al., 2023).
Analysis Note
Color: ≤ 10 Hazen
Density (d 20 °C/ 4 °C): 0.642 - 0.655
Boiling range (40-60°C): ≥ 90 % (v/v)
Aromatics (as benzene): ≤ 0.02 %
Sulfur compounds (as S): ≤ 0.005 %
Matter discolored by H₂SO₄: ≤ 10 Hazen
Iodine number: ≤ 0.3
Peroxide value (by Wheeler): ≤ 0.8
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.01 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
ACS, ISO reagent
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
<-5.8 °F
Flash Point(C)
< -21 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service