Skip to Content
Merck
All Photos(1)

Key Documents

T-007

Supelco

(±)-11-nor-9-Carboxy-Δ9-THC-D9 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19D9O4
CAS Number:
Molecular Weight:
353.50
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

OC1=C2C3C(CCC(C(O)=O)=C3)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])OC2=CC(CCCCC([2H])([2H])[2H])=C1

InChI

1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h10-12,15-16,22H,4-9H2,1-3H3,(H,23,24)/i1D3,2D3,3D3

InChI key

YOVRGSHRZRJTLZ-GQALSZNTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

11-nor-9-Carboxy-Δ9-THC, also known as THC acid, is the main secondary metabolite of Δ9-THC found in urine or blood of cannabis users. This internal standard is applicable for numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug testing and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaizheng Gong et al.
Journal of hypertension, 29(9), 1810-1819 (2011-08-13)
Pharmacological activation of peroxisome proliferator-activated receptor gamma (PPARγ) has been shown to attenuate pressure overload-induced cardiac fibrosis, suggesting that PPARγ has an antifibrotic effect. This study tested the hypothesis that there is a functional interaction between transforming growth factor-β (TGF-β)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service