Skip to Content
Merck
All Photos(1)

Key Documents

P-130

Supelco

Perphenazine solution

1 mg/mL in methanol (), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26ClN3OS
CAS Number:
Molecular Weight:
403.97
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (
)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

S1c2c(cc(cc2)Cl)N(c4c1cccc4)CCCN3CCN(CC3)CCO

InChI

1S/C21H26ClN3OS/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)27-21)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2

InChI key

RGCVKNLCSQQDEP-UHFFFAOYSA-N

General description

Perphenazine, sold under the brand name Trilafon, is a typical antipsychotic drug. Perphenazine is roughly five times as potent as chlorpromazine and is considered a medium-potency antipsychotic. This certified Snap-N-Spike; solution is suitable for use as a starting material in calibrators and controls for numerous GC/MS or LC/MS applications in forensic analysis, clinical toxicology, prescription monitoring and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rizwan M Gul et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 35(11), 2551-2556 (2017-04-05)
Crosslinking of ultra-high molecular weight polyethylene (UHMWPE) has been successfully used to improve its wear performance. Wear is a surface phenomenon and limiting crosslinking to a layer only on the surface is desirable, as crosslinking of the bulk of the
Anatoliy Markiv et al.
Journal of immunological methods, 364(1-2), 40-49 (2010-11-09)
We describe the facile generation of a stable recombinant antibody with intrinsic red fluorescent properties for qualitative and potentially quantitative immunofluorescence analysis. The REDantibody based on the X-ray crystallographic structures of the anti-sialyl-Tn antibody B72.3 and 3D model of the
Ricardo C Moraes et al.
Development (Cambridge, England), 136(9), 1423-1432 (2009-03-20)
Systemic hormones and local growth factor-mediated tissue interactions are essential for mammary gland development. Using phenotypic and transplantation analyses of mice carrying the mesenchymal dysplasia (mes) allele of patched 1 (Ptch1(mes)), we found that Ptch1(mes) homozygosity led to either complete

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service