Skip to Content
Merck
All Photos(1)

Key Documents

P-071

Supelco

Pain Management Multi-component Opiate Mixture-13 solution

100 μg/mL each component (10 μg/mL Fentanyl), ampule of 1.0 mL, certified reference material, Cerilliant®

Synonym(s):

Opiate Mixture-13 Pain Control Solution

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Buprenorphine, 100 μg/mL
Codeine, 100 μg/mL
Fentanyl, 10 μg/mL
Hydrocodone, 100 μg/mL
Hydromorphone, 100 μg/mL
Meperidine, 100 μg/mL
(±)-Methadone, 100 μg/mL
Morphine, 100 μg/mL
Naloxone, 100 μg/mL
Naltrexone, 100 μg/mL
Oxycodone, 100 μg/mL
Oxymorphone, 100 μg/mL
cis-Tramadol HCl, 100 μg/mL

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)
estupefaciente/psicótropo (Spain); Decreto Lei 15/93: Tabela IA/IIC (Portugal)

concentration

100 μg/mL each component (10 μg/mL Fentanyl)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A multi-component mix containing opiates methadone, buprenorphine, tramadol, codeine, fentanyl, hydrocodone, meperidine, morphine, naloxone, naltrexone, oxycodone, and oxymorphone. Opiates are a highly abused and addictive class of drugs prescribed for the treatment of pain. This certified solution mix is suitable for numerous applications from pain prescription monitoring, clinical toxicology, and forensic analysis to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Codeine
    Fentanyl
    Hydrocodone
    Hydromorphone

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service