Skip to Content
Merck
All Photos(1)

Key Documents

N-145

Supelco

Norbuprenorphine-D9 solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26D9NO4
CAS Number:
Molecular Weight:
422.61
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CC(O)(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])[C@](C1)([H])[C@@]([C@H]2[C@]([C@@]13[C@H]4C5)(CCN4)C6=C5C=CC(O)=C6O2)(CC3)OC

InChI key

YOYLLRBMGQRFTN-ZNBHVKAUSA-N

Application


  • Monitoring buprenorphine in treatment: Research highlighted the importance of monitoring buprenorphine and its metabolites, including norbuprenorphine, in patients undergoing medication-assisted treatment for opioid use disorder, emphasizing the utility of stable isotope-labeled standards like Norbuprenorphine-D₉ in mass spectrometry (Pesce et al., 2024).

  • Method development in mass spectrometry: A study focused on the simplified processing and rapid quantification of buprenorphine, norbuprenorphine, and their conjugates in human plasma using UPLC-MS/MS. The research underlines the significance of deuterated standards such as Norbuprenorphine-D₉ for accurate internal referencing in analytical methods (Zhao et al., 2024).

  • Substance detection in clinical settings: An investigation into the detection of buprenorphine, norbuprenorphine, and naloxone in adulterated urine samples addresses the forensic and clinical challenges in drug screening, where Norbuprenorphine-D₉ could serve as a critical analytical reference material (Furo et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service