Skip to Content
Merck
All Photos(1)

Key Documents

E-084

Supelco

(±)-N-Ethylcathinone Ephedrine Metabolite hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO · HCl
CAS Number:
Molecular Weight:
215.72
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCNC(C)C(O)C1=CC=CC=C1.Cl

InChI

1S/C11H17NO.ClH/c1-3-12-9(2)11(13)10-7-5-4-6-8-10;/h4-9,11-13H,3H2,1-2H3;1H

InChI key

DHNCTQIFOGRFJF-UHFFFAOYSA-N

General description

This bath salt cathinone analog is a primary urinary metabolite of N-Ethylcathinone. N-Ethylcathinone, also known as ethylpropion or ETH-CAT, is a synthetic cathinone and stimulant widely found in numerous bath salt blends and party pills. N-Ethylcathinone is also an active metabolite of the prodrug diethylcathinone, sold as the appetite suppressants Anorex, Nobesine, and Tenuate®. This certified solution standard is suitable for LC/MS or GC/MS N-Ethylcathinone testing applications in forensic analysis, urine drug testing, or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Tenuate is a registered trademark of Aventisub II Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service