Skip to Content
Merck
All Photos(1)

Key Documents

999973P

Avanti

16:0-18:1 Diether PG

Avanti Research - A Croda Brand 999973P, powder

Synonym(s):

1-O-hexadecanyl-2-O-(9Z-octadecenyl)-sn-glycero-3-phospho-(1′-rac-glycerol) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H84 NO8 P
CAS Number:
Molecular Weight:
738.07
UNSPSC Code:
51191904
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (999973P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999973P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC(O)CO)(OC[C@]([H])(OCCCCCCCC/C=C\CCCCCCCC)COCCCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C40H81O8P.H3N/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-46-40(38-48-49(43,44)47-36-39(42)35-41)37-45-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2;/h17,19,39-42H,3-16,18,20-38H2,1-2H3,(H,43,44);1H3/b19-17-;/t39?,40-;/m1./s1

InChI key

LOQYPPRARRCBCD-ORLXUSOXSA-N

General description

16:0-18:1 Diether PG (phosphoglycerol) is a class of glycerophospholipids that have hexadecane and octadecene chains attached to the sn-1 and sn-2 positions of the glycerol backbone respectively by ether bonds.

Biochem/physiol Actions

Ether linked phospholipids act as precursors for important signaling molecules. They contribute to 20 percent of the total phospholipids in humans. Pathological conditions arise due to genetic disorders affecting the synthesis of ether phospholipids. Ether glycerolipids act as membrane molecules and cellular mediators.

Packaging

5 mL Clear Glass Sealed Ampule (999973P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service